6-Chloro-o-toluidine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS20266G100
AS20266G25
ACTIAS20266G100EA
0
GBP
ACTIAS20266G100
ACTIAS20266G25
6-Chloro-o-toluidine
6-Chloro-o-toluidine
Formula:
C₇H₈ClN Boiling Pt: 215 °C (1013 hPa) Melting Pt: 2 °C Density: 1.152 g/cm³ (20 °C) |
MDL Number:
MFCD00007679 CAS Number: 87-63-8 EINECS: 201-759-2 UN: 3429 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...