2,3,5,6-Tetrafluorobenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (2,3,5,6-Tetrafluorophenyl)methanol

AS24706G100 AS24706G25
ACTIAS24706G100EA 0 GBP
ACTIAS24706G100 ACTIAS24706G25
2,3,5,6-Tetrafluorobenzyl alcohol
2,3,5,6-Tetrafluorobenzyl alcohol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
MDL Number: MFCD00792428
CAS Number: 4084-38-2
EINECS: 443-840-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR