2-Fluorobenzylamine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS21195G100 AS21195G25
ACTIAS21195G100EA 0 GBP
ACTIAS21195G100 ACTIAS21195G25
2-Fluorobenzylamine
2-Fluorobenzylamine
Formula: C₇H₈FN
Boiling Pt: 73 °C (1013 hPa)
Density: 1.09 g/cm³ (20 °C)
MDL Number: MFCD00008107
CAS Number: 89-99-6
EINECS: 201-957-9
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR