5-Iodo-m-xylene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS24808G100 AS24808G25
ACTIAS24808G100EA 0 GBP
ACTIAS24808G100 ACTIAS24808G25
5-Iodo-m-xylene
5-Iodo-m-xylene
Formula: C₈H₉I
Boiling Pt: 234…235 °C (1013 hPa)
Density: 1,61 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00060659
CAS Number: 22445-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR