2,3-Dichloro-α,α,α-trifluorotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24691G100 AS24691G25 AS24691G5
ACTIAS24691G100EA 0 GBP
ACTIAS24691G100 ACTIAS24691G25 ACTIAS24691G5
2,3-Dichloro-α,α,α-trifluorotoluene
2,3-Dichloro-α,α,α-trifluorotoluene
Formula: C₇H₃Cl₂F₃
MDL Number: MFCD00069115
CAS Number: 54773-19-2
EINECS: 259-336-3
UN: 2234
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR