Cyclopentylmethanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Cyclopentanemethanol

AS22199G100 AS22199G25
ACTIAS22199G100EA 0 GBP
ACTIAS22199G100 ACTIAS22199G25
Cyclopentylmethanol
Cyclopentylmethanol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 162
Density: 0.926 g/cm³ (20 °C)
MDL Number: MFCD00001384
CAS Number: 3637-61-4
EINECS: 222-861-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR