2,4-Dihydroxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Pyridine-2,4-diol, 2,4-Pyridindiol, 3-Deazauracil, 4-Hydroxy-2-pyridone

AS23087G100 AS23087G25
ACTIAS23087G100EA 0 GBP
ACTIAS23087G100 ACTIAS23087G25
2,4-Dihydroxypyridine
2,4-Dihydroxypyridine
Formula: C₅H₅NO₂
MW: 111.10 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 275…277 °C
MDL Number: MFCD00006273
CAS Number: 626-03-9
EINECS: 210-924-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR