5-Fluoro-2-nitroanisole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluoro-2-methoxy-1-nitrobenzene

AS21529G100 AS21529G25
ACTIAS21529G100EA 0 GBP
ACTIAS21529G100 ACTIAS21529G25
5-Fluoro-2-nitroanisole
5-Fluoro-2-nitroanisole
Formula: C₇H₆FNO₃
MW: 171.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077541
CAS Number: 448-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR