3-Fluoro-4-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20765G100 AS20765G25 AS20765G5
ACTIAS20765G100EA 154 GBP
ACTIAS20765G100 ACTIAS20765G25 ACTIAS20765G5
3-Fluoro-4-nitrobenzoic acid
3-Fluoro-4-nitrobenzoic acid
Formula: C₇H₄FNO₄
MW: 185.11 g/mol
MDL Number: MFCD01862092
CAS Number: 403-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR