2-Fluoro-p-toluic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21289G100 AS21289G25 AS21289G5
ACTIAS21289G100EA 264 GBP
ACTIAS21289G100 ACTIAS21289G25 ACTIAS21289G5
2-Fluoro-p-toluic acid
2-Fluoro-p-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD03092906
CAS Number: 7697-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR