5,6-Difluoroindoline-2,3-dione

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5,6-Difluoroisatin

AS23612G25 AS23612G5
ACTIAS23612G25EA 310 GBP
ACTIAS23612G25 ACTIAS23612G5
5,6-Difluoroindoline-2,3-dione
5,6-Difluoroindoline-2,3-dione
Formula: C₈H₃F₂NO₂
MW: 183.110 g/mol
MDL Number: MFCD02998377
CAS Number: 774-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR