2,3,4-Trifluorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22530G100 AS22530G25
ACTIAS22530G100EA 190 GBP
ACTIAS22530G100 ACTIAS22530G25
2,3,4-Trifluorobenzonitrile
2,3,4-Trifluorobenzonitrile
Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 116…118 °C (100 mmHg)
Density: 1.12 g/cm³ (20 °C)
MDL Number: MFCD00013288
CAS Number: 143879-80-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR