α,α,α-Trifluoro-3-tolualdehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20028G100 AS20028G25
ACTIAS20028G100EA 0 GBP
ACTIAS20028G100 ACTIAS20028G25
α,α,α-Trifluoro-3-tolualdehyde
α,α,α-Trifluoro-3-tolualdehyde
Formula: C₈H₅F₃O
Boiling Pt: 83 °C (1013 hPa)
Density: 1,3024 g/cm³ (20 °C)
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR