Indole-3-carboxylic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1H-Indole-3-carboxylic acid
AS21199G100
AS21199G25
ACTIAS21199G100EA
0
GBP
ACTIAS21199G100
ACTIAS21199G25
Indole-3-carboxylic acid
Indole-3-carboxylic acid
Formula:
C₉H₇NO₂ |
MDL Number:
MFCD00005624 CAS Number: 771-50-6 EINECS: 212-231-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...