3-Methylcinnamic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-(m-Tolyl)acrylic acid

AS21125G100 AS21125G25
ACTIAS21125G100EA 0 GBP
ACTIAS21125G100 ACTIAS21125G25
3-Methylcinnamic acid
3-Methylcinnamic acid
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 303
Melting Pt: 119…120 °C
MDL Number: MFCD00016844
CAS Number: 3029-79-6
EINECS: 221-199-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR