2',6'-Dihydroxyacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-(2,6-Dihydroxyphenyl)ethan-1-one, 1-(2,6-Dihydroxyphenyl)ethanone, 2,6-Dihydroxyacetophenone

AS20120G100 AS20120G25
ACTIAS20120G100EA 0 GBP
ACTIAS20120G100 ACTIAS20120G25
2',6'-Dihydroxyacetophenone
2',6'-Dihydroxyacetophenone
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 296
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00002270
CAS Number: 699-83-2
EINECS: 211-833-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR