3,4-Dichlorobenzyl alcohol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(3,4-Dichlorophenyl)methanol
AS22369G100
AS22369G25
ACTIAS22369G100EA
0
GBP
ACTIAS22369G100
ACTIAS22369G25
3,4-Dichlorobenzyl alcohol
3,4-Dichlorobenzyl alcohol
Formula:
C₇H₆Cl₂O MW: 177.03 g/mol Boiling Pt: 271 Melting Pt: 32…34 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004633 CAS Number: 1805-32-9 EINECS: 217-299-0 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...