2-Amino-4-chloropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20445G100 AS20445G25 AS20445G5
ACTIAS20445G100EA 0 GBP
ACTIAS20445G100 ACTIAS20445G25 ACTIAS20445G5
2-Amino-4-chloropyridine
2-Amino-4-chloropyridine
Formula: C₅H₅ClN₂
MDL Number: MFCD04113820
CAS Number: 19798-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR