2-Chloro-5-iodobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22435G100 AS22435G25
ACTIAS22435G100EA 55 GBP
ACTIAS22435G100 ACTIAS22435G25
2-Chloro-5-iodobenzoic acid
2-Chloro-5-iodobenzoic acid
Formula: C₇H₄ClIO₂
MW: 282.46 g/mol
MDL Number: MFCD00079731
CAS Number: 19094-56-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR