5,7-Dichloroisatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21324G1 AS21324G25 AS21324G5
ACTIAS21324G1EA 155 GBP
ACTIAS21324G1 ACTIAS21324G25 ACTIAS21324G5
5,7-Dichloroisatin
5,7-Dichloroisatin
Formula: C₈H₃Cl₂NO₂
MW: 216.02 g/mol
MDL Number: MFCD00043905
CAS Number: 6374-92-1
EINECS: 228-928-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR