2,4-Dichloro-6-methylpyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS24726G100 AS24726G25
ACTIAS24726G100EA 0 GBP
ACTIAS24726G100 ACTIAS24726G25
2,4-Dichloro-6-methylpyrimidine
2,4-Dichloro-6-methylpyrimidine
Formula: C₅H₄Cl₂N₂
Boiling Pt: 219 °C (1013 hPa)
Melting Pt: 47 °C
MDL Number: MFCD00006064
CAS Number: 5424-21-5
EINECS: 226-563-4
UN: 3263
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR