2,4,6-Trichloropyrimidine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS20097G100
AS20097G25
ACTIAS20097G100EA
0
GBP
ACTIAS20097G100
ACTIAS20097G25
2,4,6-Trichloropyrimidine
2,4,6-Trichloropyrimidine
Formula:
C₄HCl₃N₂ Boiling Pt: 210 °C (1013 hPa) Melting Pt: 21 °C Density: 1,595 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006063 CAS Number: 3764-01-0 EINECS: 223-183-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...