6-Chloropurine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Chloro-7H-purine, 6-Chloro-1H-purine, 6-Chloro-9H-purine

AS21493G100 AS21493G25
ACTIAS21493G100EA 0 GBP
ACTIAS21493G100 ACTIAS21493G25
6-Chloropurine
6-Chloropurine
Formula: C₅H₃ClN₄
MW: 154.56 g/mol
Boiling Pt: 185…192 °C (2 mmHg)
Melting Pt: 178…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075825
CAS Number: 87-42-3
EINECS: 201-745-6
Merck Index: 13,02180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR