5-Chloroindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Chloro-1H-indole

AS20108G25 AS20108G5
ACTIAS20108G25EA 0 GBP
ACTIAS20108G25 ACTIAS20108G5
5-Chloroindole
5-Chloroindole
Formula: C₈H₆ClN
MW: 151.6 g/mol
Melting Pt: 71…72 °C
MDL Number: MFCD00005672
CAS Number: 17422-32-1
EINECS: 241-448-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR