(3-Bromophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21295G100 AS21295G25
ACTIAS21295G100EA 0 GBP
ACTIAS21295G100 ACTIAS21295G25
(3-Bromophenyl)acetic acid
(3-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 102…103 °C
MDL Number: MFCD00004330
CAS Number: 1878-67-7
EINECS: 217-522-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR