7-Bromoisatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 7-Bromo-1H-indole-2,3-dione, 7-Bromoindoline-2,3-dione

AS20567G25 AS20567G5
ACTIAS20567G25EA 0 GBP
ACTIAS20567G25 ACTIAS20567G5
7-Bromoisatin
7-Bromoisatin
Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
MDL Number: MFCD00774354
CAS Number: 20780-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR