3-Hydroxyphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Hydroxyphenyl)metaboric acid, (3-Hydroxyphenyl)boronic acid, Dihydroxy(3-hydroxyphenyl)borane, 3-Hydroxybenzeneboronic acid

AS20989G25 AS20989G5
ACTIAS20989G25EA 0 GBP
ACTIAS20989G25 ACTIAS20989G5
3-Hydroxyphenylboronic acid
3-Hydroxyphenylboronic acid
Formula: C₆H₇BO₃
MW: 137.93 g/mol
MDL Number: MFCD01074603
CAS Number: 87199-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR