1,3,5-Trimethyl-1H-pyrazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,3,5-Trimethyl-1H-pyrazole, 1,3,5-Trimethylpyrazole
AS24164G100
AS24164G25
ACTIAS24164G100EA
297
GBP
ACTIAS24164G100
ACTIAS24164G25
1,3,5-Trimethyl-1H-pyrazole
1,3,5-Trimethyl-1H-pyrazole
Formula:
C₆H₁₀N₂ MW: 110.16 g/mol Boiling Pt: 167…170 °C (1013 hPa) Melting Pt: 30…35 °C Density: 0.9269 g/cm³ (40 °C) |
MDL Number:
MFCD00015536 CAS Number: 1072-91-9 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...