(R)-(-)-2-Methylpiperazine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21592G100
AS21592G5
ACTIAS21592G100EA
0
GBP
ACTIAS21592G100
ACTIAS21592G5
(R)-(-)-2-Methylpiperazine
(R)-(-)-2-Methylpiperazine
Formula:
C₅H₁₂N₂ Boiling Pt: 153 °C (1013 hPa) Melting Pt: 89…91 °C |
MDL Number:
MFCD00192317 CAS Number: 75336-86-6 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...