2-Phenylbenzimidazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21797G100 AS21797G25
ACTIAS21797G100EA 0 GBP
ACTIAS21797G100 ACTIAS21797G25
2-Phenylbenzimidazole
2-Phenylbenzimidazole
Formula: C₁₃H₁₀N₂
Boiling Pt: 422 °C (1013 hPa)
Melting Pt: 211 °C
MDL Number: MFCD00005592
CAS Number: 716-79-0
EINECS: 211-939-2
Merck Index: 12,07428

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR