2-Nitroresorcinol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23094G100 AS23094G25 AS23094G5
ACTIAS23094G100EA 0 GBP
ACTIAS23094G100 ACTIAS23094G25 ACTIAS23094G5
2-Nitroresorcinol
2-Nitroresorcinol
Formula: C₆H₅NO₄
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 80…84 °C
Density: 1.3526 g/cm³ (96.7 °C)
MDL Number: MFCD00007124
CAS Number: 601-89-8
EINECS: 210-010-9
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR