3-Amino-6-methylpyridazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21019G1 AS21019G5
ACTIAS21019G1EA 0 GBP
ACTIAS21019G1 ACTIAS21019G5
3-Amino-6-methylpyridazine
3-Amino-6-methylpyridazine
Formula: C₅H₇N₃
MDL Number: MFCD01569899
CAS Number: 18591-82-7
EINECS: 242-430-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR