1,2-Difluoro-3-nitrobenzene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Chloro-9-methyl-9H-purine, 2,3-Difluoronitrobenzene
AS21521G1
AS21521G25
AS21521G5
ACTIAS21521G1EA
23
GBP
ACTIAS21521G1
ACTIAS21521G25
ACTIAS21521G5
1,2-Difluoro-3-nitrobenzene
1,2-Difluoro-3-nitrobenzene
Formula:
C₆H₃F₂NO₂ MW: 159.09 g/mol |
MDL Number:
MFCD00456802 CAS Number: 6921-22-8 EINECS: 230-035-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...