2,3-Difluoro-6-nitroaniline ≥97%

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Amino-2,3-difluoro-6-nitrobenzene

AS21953G100 AS21953G25
ACTIAS21953G100EA 0 GBP
ACTIAS21953G100 ACTIAS21953G25
2,3-Difluoro-6-nitroaniline ≥97%
2,3-Difluoro-6-nitroaniline
Formula: C₆H₄F₂N₂O₂
MW: 174.11 g/mol
MDL Number: MFCD00233313
CAS Number: 211693-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR