2-Amino-5-fluoro-4-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS35103G1 AS35103G5
ACTIAS35103G1EA 0 GBP
ACTIAS35103G1 ACTIAS35103G5
2-Amino-5-fluoro-4-methylpyridine
2-Amino-5-fluoro-4-methylpyridine
Formula: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD06659479
CAS Number: 301222-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR