2-Fluoro-5-nitrobenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS24910G100
AS24910G25
ACTIAS24910G100EA
86
GBP
ACTIAS24910G100
ACTIAS24910G25
2-Fluoro-5-nitrobenzoic acid
2-Fluoro-5-nitrobenzoic acid
Formula:
C₇H₄FNO₄ MW: 185.11 g/mol |
MDL Number:
MFCD00134238 CAS Number: 7304-32-7 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...