3-Amino-4-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23456G100 AS23456G25
ACTIAS23456G100EA 0 GBP
ACTIAS23456G100 ACTIAS23456G25
3-Amino-4-fluorobenzoic acid
3-Amino-4-fluorobenzoic acid
Formula: C₇H₆FNO₂
MDL Number: MFCD00130055
CAS Number: 2365-85-7
EINECS: 219-122-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR