2'-(Trifluoromethyl)acetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: o-(Trifluoromethyl)acetophenone, 1-[2-(Trifluoromethyl)phenyl]ethan-1-one, 1-(2-(Trifluoromethyl)phenyl)ethanone

AS20270G100 AS20270G25
ACTIAS20270G100EA 0 GBP
ACTIAS20270G100 ACTIAS20270G25
2'-(Trifluoromethyl)acetophenone
2'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: 15…17 °C
Density: 1,25 g/cm³ (20 °C)
MDL Number: MFCD00000378
CAS Number: 17408-14-9
EINECS: 241-434-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR