2-(Trifluoromethoxy)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21646G100 AS21646G25
ACTIAS21646G100EA 0 GBP
ACTIAS21646G100 ACTIAS21646G25
2-(Trifluoromethoxy)benzonitrile
2-(Trifluoromethoxy)benzonitrile
Formula: C₈H₄F₃NO
Boiling Pt: 80…82 °C (15 mmHg)
MDL Number: MFCD00042409
CAS Number: 63968-85-4
EINECS: 264-584-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR