Pyridine-2,3-dicarboxylic anhydride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,3-Pyridinedicarboxylicanhydride, 2,3-Pyridinedicarboxylic anhydride, Quinolinic anhydride

AS22809G100 AS22809G25
ACTIAS22809G100EA 0 GBP
ACTIAS22809G100 ACTIAS22809G25
Pyridine-2,3-dicarboxylic anhydride
Pyridine-2,3-dicarboxylic anhydride
Formula: C₇H₃NO₃
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 146 °C
MDL Number: MFCD00005915
CAS Number: 699-98-9
EINECS: 211-834-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR