3-(3,4-Dimethoxyphenyl)propionic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,4-Dimethoxyhydrocinnamic acid, 3-(3,4-Dimethoxyphenyl)propanoic acid
AS21496G100
AS21496G25
ACTIAS21496G100EA
0
GBP
ACTIAS21496G100
ACTIAS21496G25
3-(3,4-Dimethoxyphenyl)propionic acid
3-(3,4-Dimethoxyphenyl)propionic acid
Formula:
C₁₁H₁₄O₄ Boiling Pt: 344 °C (1013 hPa) Melting Pt: 102 °C |
MDL Number:
MFCD00002774 CAS Number: 2107-70-2 EINECS: 218-288-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...