4-Nitro-o-toluic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21458G100 AS21458G25 AS21458G5
ACTIAS21458G100EA 0 GBP
ACTIAS21458G100 ACTIAS21458G25 ACTIAS21458G5
4-Nitro-o-toluic acid
4-Nitro-o-toluic acid
Formula: C₈H₇NO₄
MDL Number: MFCD00210697
CAS Number: 1975-51-5
EINECS: 217-828-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR