4-Nitro-1-indanone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Nitroindan-1-one
AS3176G1
AS3176G5
ACTIAS3176G1EA
122
GBP
ACTIAS3176G1
ACTIAS3176G5
4-Nitro-1-indanone
4-Nitro-1-indanone
Formula:
C₉H₇NO₃ MW: 177.16 g/mol |
MDL Number:
MFCD06656901 CAS Number: 24623-25-4 EINECS: 246-367-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...