2'-Aminoacetophenone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-(2-Aminophenyl)ethanone, o-Aminoacetophenone
AS20081G100
AS20081G25
ACTIAS20081G100EA
0
GBP
ACTIAS20081G100
ACTIAS20081G25
2'-Aminoacetophenone
2'-Aminoacetophenone
Formula:
C₈H₉NO MW: 135.17 g/mol Boiling Pt: 252 Melting Pt: 20 Density: 1.12 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007717 CAS Number: 551-93-9 EINECS: 209-002-8 Merck Index: 13,00412 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...