3,4-Dicyanoaniline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,4-Dicyanoaniline, 4-Aminophthalonitrile
AS23165G25
AS23165G5
ACTIAS23165G25EA
0
GBP
ACTIAS23165G25
ACTIAS23165G5
3,4-Dicyanoaniline
3,4-Dicyanoaniline
Formula:
C₈H₅N₃ Boiling Pt: 321 °C (1013 hPa) Melting Pt: 180…182 °C |
MDL Number:
MFCD00041741 CAS Number: 56765-79-8 EINECS: 260-370-6 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...