3-Amino-6-chloropyridazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Chloropyridazin-3-amine

AS50335G100 AS50335G5
ACTIAS50335G100EA 0 GBP
ACTIAS50335G100 ACTIAS50335G5
3-Amino-6-chloropyridazine
3-Amino-6-chloropyridazine
Formula: C₄H₄ClN₃
MW: 129.55 g/mol
MDL Number: MFCD00051506
CAS Number: 5469-69-2
EINECS: 226-796-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR