2-Amino-6-chloropurine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20143G100
AS20143G25
ACTIAS20143G100EA
0
GBP
ACTIAS20143G100
ACTIAS20143G25
2-Amino-6-chloropurine
2-Amino-6-chloropurine
Formula:
C₅H₄ClN₅ Melting Pt: 300 °C |
MDL Number:
MFCD00075252 CAS Number: 10310-21-1 EINECS: 233-686-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...