2-Chloro-3-iodopyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21309G25 AS21309G5
ACTIAS21309G25EA 0 GBP
ACTIAS21309G25 ACTIAS21309G5
2-Chloro-3-iodopyridine
2-Chloro-3-iodopyridine
Formula: C₅H₃ClIN
MW: 239.44 g/mol
MDL Number: MFCD00661298
CAS Number: 78607-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR