5-Chloro-1-indanone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21328G100 AS21328G25
ACTIAS21328G100EA 0 GBP
ACTIAS21328G100 ACTIAS21328G25
5-Chloro-1-indanone
5-Chloro-1-indanone
Formula: C₉H₇ClO
MW: 166.61 g/mol
Boiling Pt: 150 °C (0.2 mmHg)
Melting Pt: 100
MDL Number: MFCD00041456
CAS Number: 42348-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR