3,6-Dichloro-4-methylpyridazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS1455G100 AS1455G25 AS1455G5
ACTIAS1455G100EA 0 GBP
ACTIAS1455G100 ACTIAS1455G25 ACTIAS1455G5
3,6-Dichloro-4-methylpyridazine
3,6-Dichloro-4-methylpyridazine
Formula: C₅H₄Cl₂N₂
Boiling Pt: 130…135 °C (1 mmHg)
Melting Pt: 87 °C
MDL Number: MFCD00006465
CAS Number: 19064-64-3
EINECS: 242-794-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR